Anti-IgG Rabbit Polyclonal Antibody (DyLight® 488)

Supplier: Rockland Immunochemical
603-441-002
RL603-441-002EA 143.13 USD
RL603-441-002
Anti-IgG Rabbit Polyclonal Antibody (DyLight® 488)
Antibodies
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR