Anti-IgG Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
602-1103
RL602-1103EA 192.24 USD
RL602-1103
Anti-IgG Goat Polyclonal Antibody
Antibodies
Secondary Goat Anti-IgG F(c) Reacts with Cat (Feline)

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR