Anti-RFP Chicken Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
600-906-379S
RL600906379SEA 151.49 USD
RL600906379S
Anti-RFP Chicken Polyclonal Antibody (Biotin)
Antibodies
Type: Tag
Antigen: RFP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Chicken
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR