Anti-PRKCD Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-J29
RL600401J29EA 551.64 USD
RL600401J29
Anti-PRKCD Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PRKCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR