Anti-HDAC2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-J19
RL600401J19EA 551.64 USD
RL600401J19
Anti-HDAC2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HDAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR