Anti-HIST2H3C Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-I68
RL600401I68EA 575.67 USD
RL600401I68
Anti-HIST2H3C Rabbit Polyclonal Antibody
Antibodies
Anti-Histone H3 [Sym-dimethyl Arg8] antibody is useful for Western Blot, Chromatin Immunoprecipitation and Dot Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~15.4 kDa corresponding to Histone H3

Recommended Dilutions: ChIP: 2-5µg/million cells; Western Blot: 2µg/ml; Other Dilution: Dot Blot 1µg/ml

Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, C. elegans, Rat, Chicken, Xenopus, Drosophila, plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR