Anti-HIST2H3C Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-I66
RL600401I66EA 575.67 USD
RL600401I66
Anti-HIST2H3C Rabbit Polyclonal Antibody
Antibodies
Anti-Histone H3 [Trimethyl Lys9, p Thr6] antibody is useful for Western Blot, Immunocytochemistry, Immunofluorescence, Chromatin Immunoprecipitation, and Dot Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band appro

Recommended Dilutions: ChIP: 2-5µg/million cells; Immunofluorescence: 1:200; Western Blot: 1:500; Immunochemistry: 1:200; Other Dilution: Dot Blot 1:1000

Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, C. elegans, Rat, Chicken, Xenopus, Drosophila, plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR