Anti-S1PR4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-I18
RL600401I18EA 575.67 USD
RL600401I18
Anti-S1PR4 Rabbit Polyclonal Antibody
Antibodies
Anti-Edg6 is suitbale for use in IF/ICC and WB. Expect a band approximately 35kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: S1PR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR