Anti-SMAD3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-E70 600-401-E70S
RL600401E70EA 551.64 USD
RL600401E70 RL600401E70S
Anti-SMAD3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SMAD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR