Anti-ABCB5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-A77S
RL600401A77SEA 151.49 USD
RL600401A77S
Anti-ABCB5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ABCB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR