Anti-PDCD4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-964S
RL600401964SEA 151.49 USD
RL600401964S
Anti-PDCD4 Rabbit Polyclonal Antibody
Antibodies
Anti-Pdcd4 pS457 affinity purified antibody has been tested for use in ELISA and western blotting.  Specific conditions for reactivity should be optimized by the end user. By western blot, a band approximately 52 kDa in size.

Recommended Dilutions: ELISA: 1:400000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1.25 - 2.5 µg/ml; Other Dilution: User Optimized

Type: Primary
Antigen: PDCD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR