Anti-VSV-G Rabbit Polyclonal Antibody (DyLight® 405)

Supplier: Rockland Immunochemical
600-446-386
RL600-446-386EA 520.3 USD
RL600-446-386
Anti-VSV-G Rabbit Polyclonal Antibody (DyLight® 405)
Antibodies
Primary Rabbit Anti-VSV-G EPITOPE TAG Reacts with Pan

Type: Tag
Antigen: VSV-G
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype:
Reactivity: Many Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR