Anti-MYC Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
600-406-381
RL600-406-381EA 498.36 USD
RL600-406-381
Anti-MYC Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Tag
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR