Anti-CAV1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-J63
RL600-401-J63EA 551.64 USD
RL600-401-J63
Anti-CAV1 Rabbit Polyclonal Antibody
Antibodies
Anti-Caveolin1 antibody is useful for ELISA, Immunohistochemistry, and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~21kDa corresponding to the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1µg/ml

Type: Primary
Antigen: CAV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR