Anti-PCLO Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-G30
RL600-401-G30EA 534.54 USD
RL600-401-G30
Anti-PCLO Rabbit Polyclonal Antibody
Antibodies
Anti-Picollo Antibody is suitable for use in WB and IHC. Expect a band approximately ~550kDa corresponding to the molecular weight of Picollo. Multiple isoforms can be detected. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PCLO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR