Anti-GRM7 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-D78
RL600-401-D78EA 656.12 USD
RL600-401-D78
Anti-GRM7 Rabbit Polyclonal Antibody
Antibodies
Anti-mGluR7 pS862 (Rabbit) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 102 kDa in size corresponding to the mGluR7 Protein phosphorylated at Ser86.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GRM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR