Anti-GABRA6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-D48 600-401-D48
RL100-401-D48EA 578.81 USD
RL100-401-D48 RL600-401-D48
Anti-GABRA6 Rabbit Polyclonal Antibody
Antibodies
Anti-GABA(A) Receptor alpha 6 (Rabbit) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 57 kDa in size corresponding to the alpha 6 subunit of the GABA(A) Receptor.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GABRA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR