Anti-YWHAB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-D02
RL600-401-D02EA 656.12 USD
RL600-401-D02
Anti-YWHAB Rabbit Polyclonal Antibody
Antibodies
Anti-14-3-3 pS58 antibody is suitable for use in ELISA, Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 29 kDa in size corresponding to the 14-3-3 Proteins phosphorylated at Ser58.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000; Immunochemistry: 1:500

Type: Primary
Antigen: YWHAB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR