Anti-CTCF Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-C42 600-401-C42S
RL600-401-C42EA 551.64 USD
RL600-401-C42 RL600-401-C42S
Anti-CTCF Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody is suitable for ELISA, immunochemistry, and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 82.8 kDa or 150 kDa in size corresponding to CTCF by western blotting.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:500-1:2000; Immunochemistry: 5µg/ml;

Type: Primary
Antigen: CTCF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR