Anti-CENPU Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-A91
RL600-401-A91EA 551.64 USD
RL600-401-A91
Anti-CENPU Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CENPU
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR