Anti-SMAD2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-A59S
RL600-401-A59SEA 151.49 USD
RL600-401-A59S
Anti-SMAD2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SMAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR