Anti-SIX3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-A26 600-401-A26S
RL600-401-A26EA 503.58 USD
RL600-401-A26 RL600-401-A26S
Anti-SIX3 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA, immunochemistry and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 37 kDa in size corresponding to Six3 by western blotting.

Recommended Dilutions: ELISA: 1:10000-1:15000; Immunofluorescence: 1:200 - 1:1000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:100-1:250;

Type: Primary
Antigen: SIX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR