Anti-BBC3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-987
RL600-401-987EA 635.22 USD
RL600-401-987
Anti-BBC3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BBC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR