Anti-GYS1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-415
RL600-401-415EA 551.64 USD
RL600-401-415
Anti-GYS1 Rabbit Polyclonal Antibody
Antibodies
This phospho specific polyclonal antibody was tested by immunoblotting and ELISA.  By ELISA the antibody was found to be reactive with the phosphorylated form of the immunizing peptide and minimally reactive with the non-phosphorylated form of the immuniz

Recommended Dilutions: ELISA: 1:30,000 - 1:150000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Primary
Antigen: GYS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR