Anti-HA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-384
RL600-401-384EA 380.3 USD
RL600-401-384
Anti-HA Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-HA EPITOPE TAG Reacts with Pan

Type: Tag
Antigen: HA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR