Anti-TERT Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-252S
RL600-401-252SEA 151.49 USD
RL600-401-252S
Anti-TERT Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in immunoblotting, immunoprecipitation, and immunofluorescence microscopy.   In these assays, the antibody detects ectopically-expressed hTERT and high levels of endogenous hTERT.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Immunofluorescence: 1:500; Western Blot: 1:500; Immunochemistry: 1:500; IP 2µl per mg lysate

Type: Primary
Antigen: TERT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR