Anti-ING3 Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
600-101-283
RL600-101-283EA 551.64 USD
RL600-101-283
Anti-ING3 Goat Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA against the immunizing peptide. Specific conditions for western blotting reactivity should be optimized by the end user. Expect a band at approximately 47 kDa in size corresponding to ING3.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:200 - 1:2000;

Type: Primary
Antigen: ING3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR