Anti-GFP Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
600-101-215
RL600-101-215EA 712.54 USD
RL600-101-215
Anti-GFP Goat Polyclonal Antibody
Antibodies
Anti-GFP is designed to detect GFP and its variants. Goat Anti-GFP is ideal for western blotting, ELISA, immunochemistry and IP.

Recommended Dilutions: ELISA: 1:20000; Immunofluorescence: 1:500; Western Blot: 1:400 - 1:2000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: Yes;

Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Goat
Isotype: IgG
Reactivity: Green Fluorescent Protein
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR