Anti-CLU Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
600-101-198
RL600-101-198EA 635.22 USD
RL600-101-198
Anti-CLU Goat Polyclonal Antibody
Antibodies
Anti-apoLipoProtein antibodies have been used for indirect trapping ELISA for quantitation of antigen in serum using a standard curve, for immunoprecipitation and for western blotting for highly sensitive qualitative analysis.

Recommended Dilutions: ELISA: 1:5000 - 1:10000; Western Blot: 1:5000 - 1:10000; Immunochemistry: 1:50 - 1:200; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CLU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR