Anti-IL17A Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
212-406-B32
RL212-406-B32EA 531.79 USD
RL212-406-B32
Anti-IL17A Rabbit Polyclonal Antibody (Biotin)
Antibodies
This purified antibody has been tested for use in ELISA and western blotting. By western blot a band approximately 30.0 kDa in size corresponding to Rat IL17-A Protein is expected in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:20000-1:100000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: IL17A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR