Anti-IL17A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
212-401-B32
RL212-401-B32EA 551.64 USD
RL212-401-B32
Anti-IL17A Rabbit Polyclonal Antibody
Antibodies
This purified antibody has been tested for use in ELISA and western blotting. Reactivity is also expected in neutralizations, radioimmunoassay.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:1000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: IL17A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR