Anti-IL17F Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
210-406-B31
RL210-406-B31EA 531.79 USD
RL210-406-B31
Anti-IL17F Rabbit Polyclonal Antibody (Biotin)
Antibodies
This purified antibody has been tested for use in ELISA and western blotting. By western blot a band approximately 17.9 kDa in size corresponding to mouse IL17-F Protein is expected in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:20000-1:100000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: IL17F
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR