Anti-EBI3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
210-403-B66
RL210-403-B66EA 531.79 USD
RL210-403-B66
Anti-EBI3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
IL-27 is expressed in activated antigen presenting cells including monocytes, endothelial cells, and dendritic cells, for example mouse CD4 splenocytes. This purified antibody has been tested for use in western blotting.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:2500
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Primary
Antigen: EBI3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR