Anti-LEP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
210-401-320
RL210-401-320EA 1380.15 USD
RL210-401-320
Anti-LEP Rabbit Polyclonal Antibody
Antibodies
This IgG fraction of anti-Mouse Leptin was tested for use in ELISA and immunoblotting.  Reactivity in other immunoassays is unknown. The antibody will recognize recombinant and native 16 kDa leptin from mouse and human.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: LEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR