Anti-IL22 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
209-401-J33
RL209401J33EA 551.64 USD
RL209401J33
Anti-IL22 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR