Anti-TSLP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
209-401-H48S
RL209401H48SEA 151.49 USD
RL209401H48S
Anti-TSLP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TSLP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR