Anti-RBC Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
209-4139
RL209-4139EA 471 USD
RL209-4139
Anti-RBC Rabbit Polyclonal Antibody
Antibodies
Anti-Human Red Blood Cells Antibody are useful in immunochemistry. Specific conditions should be optimized by researcher.

Recommended Dilutions: Immunochemistry: 2.5µg/ml; Agglutination Titer 1:32 - 1:64 
Restore with deionized water (or equivalent); 5.0 ml

Type: Primary
Antigen: RBC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR