Anti-IFNK Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
209-401-C24
RL209-401-C24EA 551.64 USD
RL209-401-C24
Anti-IFNK Rabbit Polyclonal Antibody
Antibodies
This purified antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 19.9 kDa in size corresponding to the mature human IL-29 Protein by western blotting.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: IFNK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR