Anti-IL1B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
209-401-B73
RL209-401-B73EA 551.64 USD
RL209-401-B73
Anti-IL1B Rabbit Polyclonal Antibody
Antibodies
Anti-Human IL-1-beta has been tested for use in neutralizations, ELISA, radioimmunoassays, flow cytometry.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: IL1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Primate, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR