Anti-CCL20 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
209-401-B51
RL209-401-B51EA 551.64 USD
RL209-401-B51
Anti-CCL20 Rabbit Polyclonal Antibody
Antibodies
This purified antibody has been tested for use in neutralizations, ELISA and western blotting. Reactivity is also expected in radioimmunoassay.

Recommended Dilutions: ELISA: 1:20000-1:100000; Neutralization: 1:400; Western Blot: 1:500-1:2000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: CCL20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR