Anti-CD74 Mouse Monoclonal Antibody [clone: PIN.1]

Supplier: Rockland Immunochemical
209-301-F12
RL209-301-F12EA 588.97 USD
RL209-301-F12
Anti-CD74 Mouse Monoclonal Antibody [clone: PIN.1]
Antibodies
Anti-CD74 Antibody is suitable for WB, IP, Flow Cytometry, FACS, and IHC. Expect a band approximately ~33-35kDa Protein doublet corresponding to the molecular mass of the p33 and p35 forms of human CD74 on SDS PAGE immunoblots.

Recommended Dilutions: Western Blot: 1 µg/ml;

Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: PIN.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR