Anti-RXRG Mouse Monoclonal Antibody [clone: 1373]

Supplier: Rockland Immunochemical
209-301-E28
RL209-301-E28EA 659.25 USD
RL209-301-E28
Anti-RXRG Mouse Monoclonal Antibody [clone: 1373]
Antibodies
Anti-Retinoid X Receptor gamma antibody is suitable for use in Western Blotting to detect a single band at approximately 48 kDa corresponding to the RXR-gamma isotype in the appropriate cell lysate or extract.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RXRG
Clonality: Monoclonal
Clone: 1373
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR