Anti-IL17A Mouse Monoclonal Antibody [clone: 4H1524.1]

Supplier: Rockland Immunochemical
209-301-B32S
RL209-301-B32SEA 151.49 USD
RL209-301-B32S
Anti-IL17A Mouse Monoclonal Antibody [clone: 4H1524.1]
Antibodies
Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 4H1524.1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR