Anti-AKT2 Rat Monoclonal Antibody [clone: 11F6.B2]

Supplier: Rockland Immunochemical
200-501-J35
RL200501J35EA 565.22 USD
RL200501J35
Anti-AKT2 Rat Monoclonal Antibody [clone: 11F6.B2]
Antibodies
Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 11F6.B2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR