Anti-ELA1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
200-4668S
RL2004668SEA 151.49 USD
RL2004668S
Anti-ELA1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: ELA1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR