Anti-GLUD1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4158-0100 200-4158S
RL20041580100EA 551.64 USD
RL20041580100 RL2004158S
Anti-GLUD1 Rabbit Polyclonal Antibody
Antibodies
GLUTAMATE DEHYDROGENASE antibody has been assayed against 1.0 ug of Glutamate Dehydrogenase [Bovine Liver] in a standard sandwich ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Type: Primary
Antigen: GLUD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR