Anti-PhoA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4134-0100
RL20041340100EA 551.64 USD
RL20041340100
Anti-PhoA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PhoA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR