Anti-B2M Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
200-406-141S
RL200406141SEA 150.22 USD
RL200406141S
Anti-B2M Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: B2M
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR