Anti-FTH1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
200-406-090-0100
RL200406090010EA 531.79 USD
RL200406090010
Anti-FTH1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: FTH
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR