Anti-NR4A1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-H99
RL200401H99EA 614.33 USD
RL200401H99
Anti-NR4A1 Rabbit Polyclonal Antibody
Antibodies
Anti Nur77 antibody is suitable for use in ChIP, IF/ICC, IP and WB. Expect a band approximately 71kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: NR4A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR